Assay Details
Target Gene Details
Entrez Gene ID: | 74470 |
Gene Name: | centrosomal protein 72 |
Gene Aliases: |
2610029E11Rik, 4933440J22Rik |
Location: |
Chr.13:74036495-74062314 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cep72 | NM_028959.3 | 7 | 1069 | NP_083235.3 |
| AK011607.1 | BAB27729.1 | |||
| AK017134.1 | 7 | 1061 | BAB30613.2 | |
| AK034892.1 | 7 | 1062 | BAC28870.1 | |
| AK161033.1 | 3 | 394 | BAE36159.1 | |
| AK173189.1 | 1 | 2028 | BAD32467.1 | |
| BC100549.1 | 6 | 919 | AAI00550.1 |

Genomic Map