Assay Details
Target Gene Details
Entrez Gene ID: | 238123 |
Gene Name: | component of oligomeric golgi complex 5 |
Gene Aliases: |
5430405C01Rik, C87247, D18362, GOLTC1, GTC90 |
Location: |
Chr.12:31654868-31937630 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cog5 | NM_001163126.1 | NP_001156598.1 | ||
| AK030656.1 | BAC27065.1 | |||
| CA316530.1 |
Target Gene Details
Entrez Gene ID: | 73010 |
Gene Name: | G protein-coupled receptor 22 |
Gene Aliases: |
2900068K05Rik, AW061316 |
Location: |
Chr.12:31706867-31713926 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr22 | NM_175191.4 | 3 | 3534 | NP_780400.2 |
| AK034194.1 | 3 | 3488 | BAC28626.1 | |
| AK141569.1 | 3 | 3519 | ||
| BC089509.1 | 3 | 3535 | AAH89509.1 |
More Information
Set Membership: |
|

Genomic Map