Assay Details
Target Gene Details
Entrez Gene ID: | 50779 |
Gene Name: | regulator of G-protein signaling 6 |
Gene Aliases: |
- |
Location: |
Chr.12:82616176-83158815 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rgs6 | NM_015812.3 | 1 | 231 | NP_056627.1 |
AK052527.1 | 1 | 232 | BAC35026.1 | |
AK139323.1 | 1 | 345 | BAE23955.1 |
