Assay Details
Target Gene Details
Entrez Gene ID: | 72948 |
Gene Name: | tubulin polymerization promoting protein |
Gene Aliases: |
2900041A09Rik, AI849835 |
Location: |
Chr.13:74009419-74035753 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tppp | NM_182839.2 | 2 | 142 | NP_878259.1 |
| AK141543.1 | 2 | 86 | BAE24727.1 | |
| BC054803.1 | 2 | 83 | AAH54803.1 | |
| BY124320.1 | 2 | 142 |
More Information
Set Membership: |
|

Genomic Map