Assay Details
Target Gene Details
Entrez Gene ID: | 19720 |
Gene Name: | tripartite motif-containing 27 |
Gene Aliases: |
AW538890, Gm19403, Rfp |
Location: |
Chr.13:21179440-21194723 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim27 | NM_009054.3 | 1 | 186 | NP_033080.2 |
| AI530745.1 | ||||
| AK049314.1 | 1 | 160 | ||
| BQ573214.1 | 1 | 158 |
More Information
Set Membership: |
|

Genomic Map