Assay Details
Target Gene Details
Entrez Gene ID: | 56320 |
Gene Name: | drebrin 1 |
Gene Aliases: |
- |
Location: |
Chr.13:55473428-55488163 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dbn1 | NM_001177371.1 | 15 | 2520 | NP_001170842.1 |
| NM_001177372.1 | 14 | 2385 | NP_001170843.1 | |
| NM_019813.4 | 14 | 2382 | NP_062787.2 | |
| AK132060.1 | BAE20965.1 | |||
| BC006714.1 | 15 | 2330 | AAH06714.1 |
More Information
Set Membership: |
|

Genomic Map