Assay Details
Target Gene Details
Entrez Gene ID: | 12454 |
Gene Name: | cyclin K |
Gene Aliases: |
AW123198, AW413594, CPR4, CycK |
Location: |
Chr.12:108179446-108203362 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnk | NM_009832.2 | 9 | 1102 | NP_033962.2 |
| AK089386.1 | 9 | 1039 | ||
| AK146711.1 | 9 | 1039 | BAE27377.1 | |
| AK154679.1 | 9 | 1039 | BAE32760.1 | |
| AK154765.1 | 9 | 1039 | BAE32814.1 | |
| BC027297.1 | 9 | 997 | AAH27297.1 |
More Information
Set Membership: |
|

Genomic Map