Assay Details
Target Gene Details
Entrez Gene ID: | 18035 |
Gene Name: | nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, alpha |
Gene Aliases: |
AI462015, Nfkbi |
Location: |
Chr.12:55489409-55492647 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfkbia | NM_010907.2 | 6 | 1388 | NP_035037.2 |
| AI462015.1 | 1 | 193 | ||
| AK089215.1 | ||||
| AK161622.1 | 6 | 1300 |

Genomic Map