Assay Details
Target Gene Details
Entrez Gene ID: | 14538 |
Gene Name: | glucosaminyl (N-acetyl) transferase 2, I-branching enzyme |
Gene Aliases: |
5330430K10Rik, IGnT, IGnTA, IGnTB, IGnTC |
Location: |
Chr.13:40859768-40960892 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gcnt2 | NM_008105.3 | 1 | 1333 | NP_032131.2 |
| NM_023887.4 | NP_076376.3 | |||
| AB037597.1 | 1 | 724 | BAB03496.1 | |
| AK054226.1 | 1 | 848 | BAC35697.1 | |
| AK077598.1 | BAC36889.1 | |||
| AK080061.1 | 1 | 899 | ||
| AK156328.1 | BAE33677.1 | |||
| AY236873.1 | 1 | 1208 | AAO86063.1 | |
| AY236874.1 | AAO86064.1 | |||
| AY435148.1 | AAR95649.1 | |||
| AY435149.1 | 1 | 724 | AAR95650.1 | |
| BC094572.1 | AAH94572.1 | |||
| BQ892107.1 | ||||
| U68182.1 | 1 | 961 | AAB39621.1 |
More Information
Set Membership: |
|

Genomic Map