Assay Details
Target Gene Details
Entrez Gene ID: | 94066 |
Gene Name: | mitochondrial ribosomal protein L36 |
Gene Aliases: |
AI646041 |
Location: |
Chr.13:73331009-73332178 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl36 | NM_053163.1 | 2 | 353 | NP_444393.1 |
| AB049655.1 | 2 | 319 | BAB40860.1 | |
| AK075780.1 | 2 | 325 | BAC35954.1 | |
| AK131597.1 | 2 | 345 | BAE20708.1 | |
| AK131601.1 | 2 | 343 | BAE20711.1 | |
| AK131604.1 | 2 | 342 | BAE20714.1 | |
| AK160778.1 | 2 | 354 | BAE36002.1 | |
| BC009166.1 | 2 | 353 | AAH09166.1 |
More Information
Set Membership: |
|

Genomic Map