Assay Details
Target Gene Details
Entrez Gene ID: | 75901 |
Gene Name: | DCP1 decapping enzyme homolog A (S. cerevisiae) |
Gene Aliases: |
1110066A22Rik, 4930568L04Rik, AU019772, D14Ertd817e, Mitc1, SMIF |
Location: |
Chr.14:30479522-30527059 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcp1a | NM_133761.3 | 10 | 4976 | NP_598522.3 |
| AK004384.1 | 1 | 222 | ||
| AK155271.1 | 1 | 1779 |
More Information
Set Membership: |
|

Genomic Map