Assay Details
Target Gene Details
Entrez Gene ID: | 68558 |
Gene Name: | ankyrin repeat, family A (RFXANK-like), 2 |
Gene Aliases: |
1110004M18Rik, Ankra |
Location: |
Chr.13:98263076-98274754 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankra2 | NM_001271388.1 | 8 | 2348 | NP_001258317.1 |
| NM_001271389.1 | NP_001258318.1 | |||
| NM_001271390.1 | 7 | 1966 | NP_001258319.1 | |
| NM_001271391.1 | 8 | 1441 | NP_001258320.1 | |
| NM_001271392.1 | NP_001258321.1 | |||
| NM_023472.2 | 9 | 1823 | NP_075961.1 | |
| AF314031.1 | 9 | 1657 | AAK01620.1 | |
| AK003438.2 | ||||
| AK081695.1 | 7 | 1851 | BAC38296.1 | |
| AK161446.1 | ||||
| BC066113.1 | 7 | 1885 | AAH66113.1 | |
| BC079615.1 | AAH79615.1 | |||
| BE952624.1 | 1 | 115 | ||
| BF730624.1 | 1 | 95 |
More Information
Set Membership: |
|

Genomic Map