Assay Details
Target Gene Details
Entrez Gene ID: | 13361 |
Gene Name: | dihydrofolate reductase |
Gene Aliases: |
8430436I03Rik, AA607882, AI662710, AW555094 |
Location: |
Chr.13:92354783-92389053 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhfr | NM_010049.3 | 6 | 1685 | NP_034179.1 |
| AK031241.1 | 6 | 1319 | BAC27315.1 | |
| AK136212.1 | 6 | 1743 | BAE22879.1 | |
| BC005796.1 | 6 | 1257 | AAH05796.1 | |
| CB247191.1 | ||||
| L26316.1 | 6 | 1264 | AAA37523.1 |
More Information
Set Membership: |
|

Genomic Map