Assay Details
Target Gene Details
Entrez Gene ID: | 207304 |
Gene Name: | HECT domain containing 1 |
Gene Aliases: |
A630086P08Rik, AI844876, b2b327Clo, opm |
Location: |
Chr.12:51743721-51829605 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hectd1 | NM_144788.2 | 25 | 4420 | NP_659037.2 |
| AK141970.1 | 3 | 335 | BAE24902.1 | |
| AK173106.1 | 5 | 949 | BAD32384.1 |
More Information
Set Membership: |
|

Genomic Map