Assay Details
Target Gene Details
Entrez Gene ID: | 664883 |
Gene Name: | neuro-oncological ventral antigen 1 |
Gene Aliases: |
9430099M15Rik, G630039L02, Nova-1 |
Location: |
Chr.12:46694517-46819194 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nova1 | NM_021361.1 | 5 | 2697 | NP_067336.1 |
| AF232828.1 | 5 | 2631 | AAF35907.1 | |
| AK047565.1 | 1 | 1886 | BAC33088.1 |
More Information
Set Membership: |
|

Genomic Map