Assay Details
Target Gene Details
Entrez Gene ID: | 17755 |
Gene Name: | microtubule-associated protein 1B |
Gene Aliases: |
A230055D22, AI843217, LC1, MAP5, Mtap-5, Mtap1b, Mtap5 |
Location: |
Chr.13:99421464-99516602 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map1b | NM_008634.2 | 5 | 3879 | NP_032660.2 |
| BC138033.1 | 5 | 3809 | ||
| BC138034.1 | 5 | 3809 | ||
| X51396.1 | 5 | 3817 | CAA35761.1 |
More Information
Set Membership: |
|

Genomic Map