Assay Details
Target Gene Details
Entrez Gene ID: | 78754 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 15 |
Gene Aliases: |
4631401E18Rik, Galntl2, mpp-GalNAc-T15 |
Location: |
Chr.14:32028651-32062204 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galnt15 | NM_030166.3 | 1 | 777 | NP_084442.1 |
| AB175682.1 | 1 | 387 | BAD52070.1 | |
| AK019470.1 | 1 | 892 | BAB31741.1 | |
| AK030347.1 | ||||
| AK050075.1 | 1 | 274 | ||
| AK090330.1 | 1 | 471 | ||
| AK163883.1 | 1 | 652 | ||
| BC133711.1 | 1 | 777 |
More Information
Set Membership: |
|

Genomic Map