Assay Details
Target Gene Details
Entrez Gene ID: | 27052 |
Gene Name: | acyloxyacyl hydrolase |
Gene Aliases: |
4930433E13Rik |
Location: |
Chr.13:20794103-21036617 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aoah | NM_012054.4 | 21 | 2740 | NP_036184.1 |
| AA763211.1 | 1 | 153 | ||
| AK015300.1 | ||||
| AK084452.1 | 21 | 2735 | BAC39187.1 | |
| AK085343.1 | 21 | 2712 | ||
| AK133301.1 | 21 | 2681 | BAE21600.1 | |
| AK170293.1 | 20 | 2276 | BAE41693.1 | |
| AK171572.1 | 21 | 2559 | BAE42533.1 |
More Information
Set Membership: |
|

Genomic Map