Assay Details
Target Gene Details
Entrez Gene ID: | 58240 |
Gene Name: | HCLS1 binding protein 3 |
Gene Aliases: |
- |
Location: |
Chr.12:8313433-8343824 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hs1bp3 | NM_021429.3 | 7 | 2620 | NP_067404.2 |
| AK154696.1 | 7 | 2616 | ||
| BX630393.1 | 1 | 341 |

Genomic Map