Assay Details
Target Gene Details
Entrez Gene ID: | 18191 |
Gene Name: | neurexin III |
Gene Aliases: |
- |
Location: |
Chr.12:88722865-90334935 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nrxn3 | NM_001198587.3 | 18 | 4587 | NP_001185516.2 |
| NM_001252074.2 | 5 | 1802 | NP_001239003.1 | |
| NM_172544.3 | 18 | 3504 | NP_766132.2 | |
| AK032126.1 | 18 | 3496 | BAC27716.1 | |
| AK042730.1 | 5 | 1804 | BAC31344.1 | |
| AK129205.1 | 16 | 3287 | BAC98015.2 | |
| BC060719.1 | 19 | 4162 | AAH60719.1 |

Genomic Map