Assay Details
Target Gene Details
Entrez Gene ID: | 74386 |
Gene Name: | RMI1, RecQ mediated genome instability 1, homolog (S. cerevisiae) |
Gene Aliases: |
4932432N11Rik, C79893 |
Location: |
Chr.13:58402248-58411149 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rmi1 | NM_001168248.1 | 4 | 906 | NP_001161720.1 |
| NM_028904.3 | 3 | 821 | NP_083180.3 | |
| NR_031761.1 | 3 | 898 | ||
| AK016542.1 | 3 | 817 | BAB30292.1 | |
| AK032229.1 | 3 | 702 | BAC27772.1 | |
| AK079457.1 | 3 | 886 | BAC37651.1 | |
| AK139703.1 | 3 | 853 | ||
| BC037694.1 | 4 | 802 | AAH37694.1 |
More Information
Set Membership: |
|

Genomic Map