Assay Details
Target Gene Details
Entrez Gene ID: | 66143 |
Gene Name: | eukaryotic translation elongation factor 1 epsilon 1 |
Gene Aliases: |
1110003A02Rik |
Location: |
Chr.13:38645691-38659028 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eef1e1 | NM_025380.2 | 4 | 531 | NP_079656.1 |
| AK003335.1 | 4 | 514 | BAB22723.1 | |
| AK009889.1 | 4 | 503 | ||
| BC048466.1 | 4 | 513 | AAH48466.1 | |
| EF437326.1 | 2 | 134 |
More Information
Set Membership: |
|

Genomic Map