Assay Details
Target Gene Details
Entrez Gene ID: | 105193 |
Gene Name: | NHL repeat containing 1 |
Gene Aliases: |
AI505271, B230309E09Rik, EPM2B |
Location: |
Chr.13:47012557-47014850 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nhlrc1 | NM_175340.4 | 1 | 1144 | NP_780549.1 |
| AK045746.1 | 1 | 1115 | BAC32478.1 | |
| BC118951.1 | 1 | 1115 | ||
| BC118952.1 | 1 | 1113 |
More Information
Set Membership: |
|

Genomic Map