Assay Details
Target Gene Details
Entrez Gene ID: | 208366 |
Gene Name: | ribonuclease P 40 subunit |
Gene Aliases: |
D8Bwg1265e, Rnasep1 |
Location: |
Chr.13:35895104-35911565 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpp40 | NM_145938.4 | 8 | 2128 | NP_666050.3 |

Genomic Map