Assay Details
Target Gene Details
Entrez Gene ID: | 13003 |
Gene Name: | versican |
Gene Aliases: |
5430420N07Rik, 9430051N09, Cspg2, DPEAAE, NG2, PG-M, PG-M(V0), PG-M(V1), hdf |
Location: |
Chr.13:89655309-89742694 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vcan | NM_001081249.1 | 1 | 368 | NP_001074718.1 |
| NM_001134474.1 | 1 | 368 | NP_001127946.1 | |
| NM_001134475.1 | 1 | 368 | NP_001127947.1 | |
| NM_019389.2 | 1 | 368 | NP_062262.2 | |
| NM_172955.1 | 1 | 368 | NP_766543.1 | |
| AK014525.1 | 1 | 179 | BAB29411.3 | |
| AK032794.1 | 1 | 369 | ||
| AK034871.1 | 1 | 175 | BAC28862.1 | |
| AK135324.1 | 1 | 365 | BAE22489.1 | |
| AK163927.1 | 1 | 225 | BAE37538.1 | |
| AK165069.1 | 1 | 171 | BAE38022.1 | |
| BC021652.1 | 1 | 208 | ||
| BC096495.1 | 1 | 178 | AAH96495.1 | |
| BQ828249.1 | 1 | 231 | ||
| D28599.1 | 1 | 100 | ||
| D32040.1 | 1 | 100 | BAA06802.1 |
More Information
Set Membership: |
|

Genomic Map