Assay Details
Target Gene Details
Entrez Gene ID: | 71704 |
Gene Name: | Rho guanine nucleotide exchange factor (GEF) 3 |
Gene Aliases: |
1200004I24Rik, 9830169H03Rik, C76747, Xpln |
Location: |
Chr.14:27114823-27403916 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgef3 | NM_001289686.1 | 13 | 3009 | NP_001276615.1 |
| NM_001289687.1 | 10 | 2880 | NP_001276616.1 | |
| NM_001289688.1 | 10 | 3054 | NP_001276617.1 | |
| NM_027871.2 | 11 | 2854 | NP_082147.1 | |
| AK004600.2 | 11 | 2825 | BAB23401.1 | |
| AK029872.1 | 14 | 3039 | BAC26652.1 | |
| BC005517.1 | 10 | 2851 | AAH05517.1 | |
| BC007153.1 | 4 | 2098 | AAH07153.1 | |
| BC012262.1 | 10 | 2832 | AAH12262.1 |
More Information
Set Membership: |
|

Genomic Map