Assay Details
Target Gene Details
Entrez Gene ID: | 239364 |
Gene Name: | testis-specific protein, Y-encoded-like 5 |
Gene Aliases: |
6330530B20, E130308C19Rik |
Location: |
Chr.15:33683875-33687883 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tspyl5 | NM_001085421.1 | 1 | 2011 | NP_001078890.1 |
| AK031988.1 | 1 | 1721 | ||
| AK087510.1 | 1 | 1947 | ||
| AK173253.1 | 1 | 1954 | BAD32531.1 |
More Information
Set Membership: |
|

Genomic Map