Assay Details
Target Gene Details
Entrez Gene ID: | 67515 |
Gene Name: | tetratricopeptide repeat domain 33 |
Gene Aliases: |
2410099M07Rik, 2900001O04Rik, AI507072, AW538342, Osrf |
Location: |
Chr.15:5185485-5218336 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc33 | NM_026213.3 | 5 | 1637 | NP_080489.1 |
| AK010758.1 | 5 | 1456 | ||
| AK013464.1 | 5 | 1483 | BAB28868.1 | |
| AK050426.1 | 5 | 1477 | BAC34250.1 | |
| AK075643.1 | BAC35877.1 | |||
| BC017533.1 | 4 | 1402 | AAH17533.1 | |
| BC061018.1 | 5 | 1637 | AAH61018.1 |

Genomic Map