Assay Details
Target Gene Details
Entrez Gene ID: | 210126 |
Gene Name: | LIM domain containing preferred translocation partner in lipoma |
Gene Aliases: |
9430020K16Rik, AA959454, AU024130, B130055L10Rik, C79715, D630048H16 |
Location: |
Chr.16:24393350-24992578 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lpp | NM_001145952.1 | 12 | 8750 | NP_001139424.1 |
| NM_001145954.1 | 8 | 8164 | NP_001139426.1 | |
| NM_178665.5 | 13 | 8957 | NP_848780.3 | |
| AK050035.1 | 1 | 3568 |
More Information
Set Membership: |
|

Genomic Map