Assay Details
Target Gene Details
Entrez Gene ID: | 207781 |
Gene Name: | C2 calcium-dependent domain containing 2 |
Gene Aliases: |
5730563M15, 5830404H04Rik, AI314146, AI449533, BF168443, ORF25, mKIAA4045 |
Location: |
Chr.16:97855209-97922651 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C2cd2 | NM_174847.2 | 13 | 2022 | NP_777272.2 |
| AF416778.1 | ||||
| AK077731.1 | 13 | 1983 | ||
| AK080956.1 | 13 | 1966 | ||
| AK086520.1 | 13 | 2025 | ||
| AK143875.1 | BAE25577.1 | |||
| AK169902.1 | BAE41447.1 | |||
| AK220454.1 | 13 | 2007 | BAD90483.1 | |
| BC038064.1 | 3 | 454 | AAH38064.1 | |
| BC043688.1 | 13 | 1983 | AAH43688.1 |

Genomic Map