Assay Details
Target Gene Details
Entrez Gene ID: | 73047 |
Gene Name: | calcium/calmodulin-dependent protein kinase II inhibitor 2 |
Gene Aliases: |
2900075A18Rik |
Location: |
Chr.16:20619215-20621278 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Camk2n2 | NM_028420.2 | 2 | 942 | NP_082696.1 |
| AK013788.1 | 2 | 889 | BAB28995.1 | |
| AW120861.1 | ||||
| BC147550.1 | 2 | 919 | ||
| BC147736.2 | 4 | 827 | ||
| BC147745.1 | 3 | 859 | ||
| BC157896.1 | 2 | 919 |
Target Gene Details
Entrez Gene ID: | 107522 |
Gene Name: | endothelin converting enzyme 2 |
Gene Aliases: |
1810009K13Rik, 6330509A19Rik, 9630025D12Rik, BB127715 |
Location: |
Chr.16:20611601-20646480 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ece2 | NM_177940.1 | NP_808809.1 | ||
| NM_177941.1 | NP_808810.1 | |||
| AF489569.1 | AAO72356.1 | |||
| AF489570.1 | AAO72357.1 |
More Information
Set Membership: |
|

Genomic Map