Assay Details
Target Gene Details
Entrez Gene ID: | 13837 |
Gene Name: | Eph receptor A3 |
Gene Aliases: |
AW492086, Cek4, ETK1, End3, Hek, Hek4, Mek4, Tyro4 |
Location: |
Chr.16:63545218-63864157 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epha3 | NM_010140.3 | 17 | 3316 | NP_034270.1 |
| AK044936.1 | 1 | 344 | ||
| AK045219.1 | 17 | 3317 | BAC32266.1 | |
| AK084921.1 | 17 | 3296 | BAC39310.1 | |
| AK132335.1 | 17 | 3265 | BAE21112.1 |
More Information
Set Membership: |
|

Genomic Map