Assay Details
Target Gene Details
Entrez Gene ID: | 15557 |
Gene Name: | 5-hydroxytryptamine (serotonin) receptor 1F |
Gene Aliases: |
Htr1eb |
Location: |
Chr.16:64922423-65105784 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Htr1f | NM_008310.3 | 3 | 646 | NP_032336.1 |
| AK045364.1 | 3 | 715 | BAC32328.1 | |
| BC120507.1 | 3 | 502 | ||
| BC137604.1 | 3 | 502 | ||
| Z14224.1 | 3 | 646 | CAA78593.1 |
More Information
Set Membership: |
|

Genomic Map