Assay Details
Target Gene Details
Entrez Gene ID: | 75284 |
Gene Name: | BCDIN3 domain containing |
Gene Aliases: |
4930556P03Rik, AV138748 |
Location: |
Chr.15:99470084-99474730 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcdin3d | NM_029236.2 | 1 | 94 | NP_083512.2 |
| AK016154.1 | 1 | 95 | BAB30128.1 | |
| BC016225.1 | AAH16225.1 | |||
| BY262678.1 | 1 | 95 | ||
| CO427236.1 |

Genomic Map