Assay Details
Target Gene Details
Entrez Gene ID: | 68070 |
Gene Name: | PDZ domain containing 2 |
Gene Aliases: |
4930537L06Rik, A930022H17Rik, Gm82, Pdzk3, mKIAA0300 |
Location: |
Chr.15:12357054-12739973 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdzd2 | NM_001081064.1 | 2 | 572 | NP_001074533.1 |
| AK020883.1 | 3 | 169 | BAB32241.3 | |
| AK021207.1 | 2 | 741 | ||
| AK159017.1 | 4 | 1330 | BAE34770.1 | |
| BC115886.1 |

Genomic Map