Assay Details
Target Gene Details
Entrez Gene ID: | 268890 |
Gene Name: | limbic system-associated membrane protein |
Gene Aliases: |
5430428I19, AW046674, B130007O04Rik, D930023J12Rik, Lam, Lamp |
Location: |
Chr.16:41532788-42181679 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lsamp | NM_175548.3 | 6 | 1643 | NP_780757.1 |
| AK030681.1 | ||||
| AK044845.1 | 6 | 1643 | BAC32117.1 | |
| AK140043.1 | ||||
| AK158702.1 | BAE34618.1 |
More Information
Set Membership: |
|

Genomic Map