Assay Details
Target Gene Details
Entrez Gene ID: | 224092 |
Gene Name: | large subunit GTPase 1 homolog (S. cerevisiae) |
Gene Aliases: |
5830465I20, AA409273, D16Bwg1547e |
Location: |
Chr.16:30561369-30587589 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lsg1 | NM_178069.5 | 8 | 931 | NP_835170.1 |
| AK030940.1 | 8 | 906 | ||
| AK078501.1 | 8 | 904 | ||
| AK132527.1 | 9 | 1134 | ||
| AK145186.1 | 8 | 906 | BAE26280.1 | |
| AK146895.1 | 8 | 906 | BAE27512.1 | |
| AK186672.1 | ||||
| BC043724.1 | 8 | 931 | AAH43724.1 | |
| BC080306.1 | 6 | 662 | AAH80306.1 |

Genomic Map