Assay Details
Target Gene Details
Entrez Gene ID: | 106347 |
Gene Name: | immunoglobulin-like domain containing receptor 1 |
Gene Aliases: |
AU041483, AU044638 |
Location: |
Chr.16:36693978-36726804 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ildr1 | NM_001285788.1 | 8 | 2367 | NP_001272717.1 |
| NM_001285791.1 | 7 | 2235 | NP_001272720.1 | |
| NM_134109.2 | 8 | 2326 | NP_598870.1 | |
| AK036844.1 | 8 | 2252 | BAC29604.1 | |
| AK136284.1 | 8 | 2367 | BAE22913.1 | |
| BC010485.1 | 8 | 2266 | AAH10485.1 | |
| BC057644.1 | 7 | 2232 | AAH57644.1 |
More Information
Set Membership: |
|

Genomic Map