Assay Details
Target Gene Details
Entrez Gene ID: | 75880 |
Gene Name: | RIKEN cDNA 4930592A05 gene |
Gene Aliases: |
- |
Location: |
Chr.15:33593881-33653436 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930592A05Rik | NR_045070.1 | |||
| AK016384.1 |
Target Gene Details
Entrez Gene ID: | 54381 |
Gene Name: | carboxypeptidase Q |
Gene Aliases: |
1190003P12Rik, 2610034C17Rik, Hls2, Lal-1, Pgcp |
Location: |
Chr.15:33083129-33594552 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpq | NM_018755.2 | 9 | 1777 | NP_061225.2 |
| NM_176073.4 | 8 | 1687 | NP_788262.1 | |
| AF009513.1 | 8 | 1532 | AAC17945.1 | |
| AK032972.1 | 8 | 1552 | BAC28105.1 | |
| AK075686.1 | 8 | 1618 | BAC35891.1 | |
| AK135096.1 | 8 | 1674 | BAE22419.1 | |
| AK137164.1 | 8 | 1561 | BAE23259.1 | |
| AK152139.1 | 9 | 1655 | BAE30978.1 | |
| AK152145.1 | 9 | 1655 | BAE30982.1 | |
| BB472698.2 | 1 | 57 | ||
| BC037067.1 | 8 | 1534 | AAH37067.1 |
More Information
Set Membership: |
|

Genomic Map