Assay Details
Target Gene Details
Entrez Gene ID: | 26413 |
Gene Name: | mitogen-activated protein kinase 1 |
Gene Aliases: |
9030612K14Rik, AA407128, AU018647, C78273, ERK, Erk2, MAPK2, PRKM2, Prkm1, p41mapk, p42mapk |
Location: |
Chr.16:16983382-17047453 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapk1 | NM_011949.3 | 9 | 2580 | NP_036079.1 |
| AK035386.1 | 9 | 2554 | BAC29053.1 | |
| AK048159.1 | 1 | 838 | ||
| AK087925.1 | 9 | 2555 | BAC40044.1 | |
| AK132241.1 | 9 | 2568 | BAE21053.1 | |
| AK148853.1 | 9 | 2542 | BAE28679.1 | |
| BC006708.1 | 4 | 1626 | AAH06708.2 | |
| BC058258.1 | 9 | 2523 | AAH58258.1 |
More Information
Set Membership: |
|

Genomic Map