Assay Details
Target Gene Details
Entrez Gene ID: | 67501 |
Gene Name: | coiled-coil domain containing 50 |
Gene Aliases: |
C3orf6 |
Location: |
Chr.16:27388765-27452218 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc50 | NM_001025615.3 | 10 | 4965 | NP_001020786.1 |
| NM_001289436.1 | 12 | 5056 | NP_001276365.1 | |
| NM_026202.4 | 11 | 5010 | NP_080478.2 | |
| AK162970.1 | 1 | 829 |

Genomic Map