Assay Details
Target Gene Details
Entrez Gene ID: | 20618 |
Gene Name: | synuclein, gamma |
Gene Aliases: |
C79089, persyn |
Location: |
Chr.14:34370274-34374669 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sncg | NM_011430.3 | 5 | 683 | NP_035560.1 |
| AF017255.1 | 5 | 662 | AAC98893.1 | |
| AK141509.1 | 5 | 667 | BAE24706.1 | |
| BC028508.1 | 5 | 674 | AAH28508.1 |
More Information
Set Membership: |
|

Genomic Map