Assay Details
Target Gene Details
Entrez Gene ID: | 378435 |
Gene Name: | v-maf musculoaponeurotic fibrosarcoma oncogene family, protein A (avian) |
Gene Aliases: |
RIPE3b1 |
Location: |
Chr.15:75746843-75747922 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mafa | NM_194350.1 | 1 | 902 | NP_919331.1 |
More Information
Set Membership: |
|

Genomic Map