Assay Details
Target Gene Details
Entrez Gene ID: | 73261 |
Gene Name: | RIKEN cDNA 1700037C18 gene |
Gene Aliases: |
2810022L20Rik |
Location: |
Chr.16:3905798-3908690 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700037C18Rik | NM_028484.2 | 3 | 847 | NP_082760.2 |
| AK012792.1 | 3 | 818 | BAC25379.1 | |
| BC147295.1 | 3 | 635 | ||
| CR517846.1 |

Genomic Map