Assay Details
Target Gene Details
Entrez Gene ID: | 223227 |
Gene Name: | SRY (sex determining region Y)-box 21 |
Gene Aliases: |
Sox25 |
Location: |
Chr.14:118233233-118237030 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sox21 | NM_177753.3 | 1 | 2530 | NP_808421.1 |
| AY069926.1 | 1 | 1348 | AAL49967.1 | |
| AY142959.1 | 1 | 1402 | AAN60055.1 | |
| BC063054.1 | 2 | 1318 | AAH63054.1 | |
| BC069176.1 | 2 | 1831 | AAH69176.1 |
More Information
Set Membership: |
|

Genomic Map