Assay Details
Target Gene Details
Entrez Gene ID: | 380977 |
Gene Name: | RIKEN cDNA A330009N23 gene |
Gene Aliases: |
- |
Location: |
Chr.15:101221191-101225186 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A330009N23Rik | NR_045327.1 | |||
| NR_045328.1 | ||||
| AK039268.1 | ||||
| BY254676.1 | ||||
| BY777609.1 |
Target Gene Details
Entrez Gene ID: | 56149 |
Gene Name: | GRP1 (general receptor for phosphoinositides 1)-associated scaffold protein |
Gene Aliases: |
tamalin |
Location: |
Chr.15:101224207-101232756 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grasp | NM_019518.3 | 1 | 283 | NP_062391.3 |
| AB041603.1 | 1 | 281 | BAA95086.1 | |
| AF236099.1 | 1 | 246 | AAF36997.1 | |
| BC046307.1 | 1 | 251 | AAH46307.1 | |
| BY730445.1 | 1 | 302 |
More Information
Set Membership: |
|

Genomic Map