Assay Details
Target Gene Details
Entrez Gene ID: | 116939 |
Gene Name: | patatin-like phospholipase domain containing 3 |
Gene Aliases: |
Adpn |
Location: |
Chr.15:84167783-84189521 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnpla3 | NM_054088.3 | 8 | 1696 | NP_473429.2 |
| AK158566.1 | ||||
| AY037763.1 | AAK68636.1 | |||
| BC028792.1 | AAH28792.1 |
More Information
Set Membership: |
|

Genomic Map