Assay Details
Target Gene Details
Entrez Gene ID: | 67394 |
Gene Name: | RIKEN cDNA 4930404I05 gene |
Gene Aliases: |
- |
Location: |
Chr.16:91011249-91016503 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930404I05Rik | NR_028368.1 | |||
| AK006191.1 | ||||
| AK015081.1 | ||||
| AK029496.1 | 1 | 507 |
Target Gene Details
Entrez Gene ID: | 67367 |
Gene Name: | PAX3 and PAX7 binding protein 1 |
Gene Aliases: |
1810007M14Rik, C21orf66, Gcfc, Gcfc1, Pax3/7bp |
Location: |
Chr.16:91014037-91044734 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Paxbp1 | NM_026110.2 | 18 | 3619 | NP_080386.3 |
More Information
Set Membership: |
|

Genomic Map