Assay Details
Target Gene Details
Entrez Gene ID: | 12294 |
Gene Name: | calcium channel, voltage-dependent, alpha2/delta subunit 3 |
Gene Aliases: |
Cacnad3 |
Location: |
Chr.14:28904943-29721864 on Build GRCm38 |
Assay Gene Location: | Within Intron 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacna2d3 | NM_009785.1 | NP_033915.1 | ||
| AJ010949.1 | CAA09423.1 |
Target Gene Details
Entrez Gene ID: | 319476 |
Gene Name: | leucine-rich repeats and transmembrane domains 1 |
Gene Aliases: |
A930016D02Rik |
Location: |
Chr.14:29018208-29033642 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrtm1 | NM_176920.4 | 2 | 897 | NP_795894.1 |
| AK044492.1 | 2 | 851 | BAC31949.1 | |
| AK138618.1 | 2 | 715 | BAE23722.1 | |
| BC099587.1 | 2 | 695 | AAH99587.1 | |
| BC132594.1 | 2 | 601 | ||
| BC145990.1 | 2 | 601 |
More Information
Set Membership: |
|

Genomic Map