Assay Details
Target Gene Details
Entrez Gene ID: | 16995 |
Gene Name: | leukotriene B4 receptor 1 |
Gene Aliases: |
BLT1, BLTR, Ltb4r, mBLTR |
Location: |
Chr.14:55765050-55768494 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ltb4r1 | NM_008519.2 | 2 | 1179 | NP_032545.1 |
| AF044030.1 | 2 | 1175 | AAC61677.1 | |
| AF110104.1 | 2 | 1126 | AAD09817.1 | |
| AK010895.1 | 2 | 1164 | ||
| AK155181.1 | 3 | 972 | BAE33098.1 | |
| BC064063.1 | 2 | 1110 | AAH64063.1 |
More Information
Set Membership: |
|

Genomic Map